Anti-RGS13 Goat Polyclonal Antibody

Lieferant: ProSci
PRSI46-973EA 420 EUR
PRSI46-973
Anti-RGS13 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32000. Approximately 19 kDa band observed in human Peripheral Blood Mononucleocyte lysates (calculated MW of 19.1 kDa according to NP_658912.1). Recommended concentration: 0.5-1.5 μg/mL.

RGS13 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

RGS13 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: RGS13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR