Anti-COG1 Goat Polyclonal Antibody

Lieferant: ProSci
PRSI46-962EA 420 EUR
PRSI46-962
Anti-COG1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:64000. Approximately 110 kDa band observed in human brain (cerebellum, amygdala) lysates (calculated MW of 109 kDa according to NP_061184.1). Recommended concentration: 0.1-0.3 μg/mL. an additional band of unknown identity was also consistently observed at 45 kDa . This band was successfully blocked by incubation with the immunizing peptide.

COG1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

COG1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: COG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR