Anti-Dpm2 Goat Polyclonal Antibody

Lieferant: ProSci
PRSI46-961EA 345 EUR
PRSI46-961
Anti-Dpm2 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:16000. Approximately 25 kDa band in mouse eye lysates after 0.02 μg/mL antibody staining.4 kDa according to NP_034203.1. The 25 kDa band was successfully blocked by incubation with the immunizing peptide.

DPM2 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

DPM2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: Dpm2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR