Anti-GRN Goat Polyclonal Antibody

Lieferant: ProSci
PRSI46-960EA 420 EUR
PRSI46-960
Anti-GRN Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:8000. Approximately 60 kDa band observed in lysates of A549 and HeLa (calculated MW of 63.5 kDa according to NP_002078.1). Recommended concentration: 1-3 μg/mL.

Granulin antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Granulin antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GRN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR