Anti-TFAP2D Goat Polyclonal Antibody

Lieferant: ProSci
PRSI46-957EA 345 EUR
PRSI46-957
Anti-TFAP2D Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:64000.

TFAP2D antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

TFAP2D antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TFAP2D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR