Anti-NAT2 Goat Polyclonal Antibody

Lieferant: ProSci
PRSI46-943EA 435 EUR
PRSI46-943
Anti-NAT2 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:16000. Approximately 27 kDa band in human kidney, liver and testis lysates after 1 μg/mL antibody staining.6 kDa according to NP_000006.2. The 27 kDa band was successfully blocked by incubation with the immunizing peptide.

NAT2 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

NAT2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: NAT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR