Anti-CHRNB2 Goat Polyclonal Antibody

Lieferant: ProSci
PRSI46-942EA 420 EUR
PRSI46-942
Anti-CHRNB2 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:64000. Approximately 48 kDa band observed in human brain (cerebellum, cerebral cortex) lysates (calculated MW of 57.0 kDa according to NP_000739.1). The observed molecular weight corresponds to earlier findings with different antibodies from other commercial sources. Recommended concentration: 1-3 μg/mL.

CHRNB2 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

CHRNB2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CHRNB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR