Anti-TRPM8 Goat Polyclonal Antibody

Lieferant: ProSci
PRSI46-940EA 345 EUR
PRSI46-940
Anti-TRPM8 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:8000. Approximately 28 kDa band in lysates of cell line NIH3T3 after 1 μg/mL antibody staining. The 28 kDa band was successfully blocked by incubation with the immunizing peptide.

TRPM8 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

TRPM8 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TRPM8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR