Anti-Tankyrase 2 Goat Polyclonal Antibody

Lieferant: ProSci
PRSI46-937EA 553 EUR
PRSI46-937
Anti-Tankyrase 2 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:16000. Approximately 125 kDa band observed in Jurkat lysate (calculated MW of 127 kDa according to NP_079511.1). Recommended concentration: 0.5-1.5 μg/mL. an additional band of unknown identity was also consistently observed at 45 kDa . This band was successfully blocked by incubation with the immunizing peptide.

Tankyrase 2 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Tankyrase 2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: Tankyrase 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR