Anti-RNF139 Goat Polyclonal Antibody

Lieferant: ProSci
PRSI46-933EA 420 EUR
PRSI46-933
Anti-RNF139 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:16000. Experiments gave bands at approximately 150 kDa and 50 kDa in human skeletal Mucle lysate after 0.5 μg/mL antibody staining. This protein has a calculated MW of 76.0 kDa according to NP_009149.2.The observed molecular weight corresponds to earlier findings with different antibodies from other commercial sources. Recommended concentration: 0.5-1.5 μg/mL.

TRC8 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

TRC8 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: RNF139
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR