Anti-TRPC4 Goat Polyclonal Antibody

Lieferant: ProSci
PRSI46-913EA 420 EUR
PRSI46-913
Anti-TRPC4 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 110 kDa band observed in rat brain lysates (calculated MW of 112 kDa according to human NP_057263.1 and to rat NP_536321.1). Recommended concentration: 0.3-1 μg/mL.

TRPC4 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

TRPC4 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TRPC4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR