Anti-TMPRSS4 Goat Polyclonal Antibody

Lieferant: ProSci
PRSI46-909EA 553 EUR
PRSI46-909
Anti-TMPRSS4 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:1,000. Western Blot: Approximately 48 kDa band observed in human bone marrow lysates (calculated MW of 48.2 kDa according to NP_063947.1 and of 47.6 kDa according to NP_001077416.1). Recommended concentration: 1-3

TMPRSS4 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

TMPRSS4 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TMPRSS4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR