Anti-TFEB Goat Polyclonal Antibody

Lieferant: ProSci
PRSI46-906EA 553 EUR
PRSI46-906
Anti-TFEB Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 55 kDa band observed in human lung lysates (calculated MW of 52.9 kDa according to NP_009093.1). Recommended concentration: 1-3 μg/mL.

Transcription Factor EB antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Transcription Factor EB antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TFEB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR