Anti-SNX19 Goat Polyclonal Antibody

Lieferant: ProSci
PRSI46-900EA 420 EUR
PRSI46-900
Anti-SNX19 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:16,000. Western Blot: Approximately110 kDa band observed in human skeletal muscle lysates (calculated MW of 109 kDa according to NP_055573.2). Recommended concentration: 0.3-1 μg/mL.

Sorting Nexin 19 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Sorting Nexin 19 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SNX19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR