Anti-PALLD Goat Polyclonal Antibody

Lieferant: ProSci
PRSI46-884EA 345 EUR
PRSI46-884
Anti-PALLD Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:2,000. Western Blot: Preliminary experiments in human liver, heart and ovary lysates gave no specific signal but low background (at antibody concentration up to 1 μg/mL). We would appreciate any feed

Palladin antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Palladin antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PALLD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR