Anti-D130043K22Rik Goat Polyclonal Antibody

Lieferant: ProSci
PRSI46-866EA 420 EUR
PRSI46-866
Anti-D130043K22Rik Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:128,000. Western Blot: Approximately 150 kDa band observed in mouse brain lysates (calculated MW of 118 kDa according to mouse NP_001074520.1). The observed molecular weight corresponds to earlier findings in li

KIAA0319 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

KIAA0319 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: D130043K22Rik
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR