Anti-GDF2 Goat Polyclonal Antibody

Lieferant: ProSci
PRSI46-848EA 435 EUR
PRSI46-848
Anti-GDF2 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:4,000. Western Blot: Approximately 27 kDa band in human bone marrow, liver, and cerebellum lysates after 1 μg/mL antibody staining. Please note that currently we canno

GDF2 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

GDF2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GDF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR