Anti-FZD2 Goat Polyclonal Antibody

Lieferant: ProSci
PRSI46-844EA 420 EUR
PRSI46-844
Anti-FZD2 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:64,000. Western Blot: Approximately 23 kDa band in human colon, heart and ovary lysates after 0.1 μg/mL antibody staining. Immunohistochemistry: In paraffin embedded human thyroid gland shows cytoplasm and membranous staining in epithelial cells of the follicles. Recommended concentration, 2-5 μg/mL.

Frizzled 2 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Frizzled 2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: FZD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR