Anti-FOXI2 Goat Polyclonal Antibody

Lieferant: ProSci
PRSI46-843EA 345 EUR
PRSI46-843
Anti-FOXI2 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:8,000.

Forkhead Box I2 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Forkhead Box I2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: Foxi2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR