Anti-SH2D1B Goat Polyclonal Antibody

Lieferant: ProSci
PRSI46-835EA 420 EUR
PRSI46-835
Anti-SH2D1B Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:128,000. Western Blot: Approximately 14 kDa band observed in lysates of cell line A431 (calculated MW of 16.6 kDa according to NP_002643.1). Recommended concentration: 0.3-1 μg/mL.

EAT2 Phospho Y127 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

EAT2 pH ospH o Y127 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SH2D1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR