Anti-DYX1C1 Goat Polyclonal Antibody

Lieferant: ProSci
PRSI46-834EA 420 EUR
PRSI46-834
Anti-DYX1C1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 48+40 kDa bands observed in human brain (cerebral cortex and frontal cortex) lysates (calculated MW of 48.5 kDa according to NP_570722.2 and of 44.0 kDa according to NP_001028731

DYX1C1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

DYX1C1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: DYX1C1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR