Anti-DMP1 Goat Polyclonal Antibody

Lieferant: ProSci
PRSI46-832EA 420 EUR
PRSI46-832
Anti-DMP1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:4,000. Western Blot: Approximately 55 kDa band observed in human Ileum and kidney lysates (calculated MW of 55.8 kDa according to NP_004398.1 and of 54.1 according to NP_001073380.1). Recommended concentration: 1

DMP1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

DMP1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: DMP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR