Anti-DAGLA Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-829EA 420 EUR
PRSI46-829
Anti-DAGLA Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:128,000. Western Blot: Approximately 125 kDa band observed in human liver lysates (calculated MW of 115 kDa according to NP_006124.1). Recommended concentration: 0.3-1 μg/mL. Some minor background is detected.

DAGLA antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

DAGLA antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: DAGLA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR