Anti-ATG16L1 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-814EA 420 EUR
PRSI46-814
Anti-ATG16L1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 65 kDa band observed in mouse brain lysates (calculated MW of 68.1 kDa according to mouse NP_084122.2). Recommended concentration: 0.01-0.1 μg/mL.

ATG16L1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

ATG16L1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ATG16L1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR