Anti-ABHD12 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-799EA 420 EUR
PRSI46-799
Anti-ABHD12 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 40 kDa band observed in mouse brain and rat brain lysates (calculated MW of 45.1 kDa according to human NP_001035937.1 and of 45.3 kDa according to mouse NP_077785.2). Recommend

ABHD12 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

ABHD12 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ABHD12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR