Anti-ABCB8 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-793EA 345 EUR
PRSI46-793
Anti-ABCB8 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:128,000. Western Blot: Approximately 22 kDa band in lysates of cell lines HeLa and Jurkat after 0.5 μg/mL antibody staining. Please note that currently we cannot find an explan

ABCB8 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

ABCB8 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ABCB8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR