Anti-ABCA6 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-790EA 345 EUR
PRSI46-790
Anti-ABCA6 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 50 kDa band in human brain (cerebellum, cerebral cortex, frontal cortex) lysates after 0.5 μg/mL antibody staining. Please note that current

ABCA6 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

ABCA6 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ABCA6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR