Anti-ABCA2 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-787EA 553 EUR
PRSI46-787
Anti-ABCA2 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:2,000. Western Blot: Approximately 35 kDa band in human brain (amygdala, frontal cortex, hippocampus and Substantia Nigra) lysates after 0.3 μg/mL antibody staining. Please no

ABCA2 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

ABCA2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ABCA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR