Anti-ANXA1 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-785EA 553 EUR
PRSI46-785
Anti-ANXA1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 35 kDa band observed in human lysates of cell lines A431 and K562 (calculated MW of 38.7 kDa according to NP_000691.1). The observed molecular weight corresponds to earlier findings in literature with different antibodies (Paweletz et al, tumor Res. 2000 Nov15;60 (22) :6293-7. PMID: 11103786). Recommended concentration: 0.5-1.5 μg/mL.

Annexin A1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Annexin A1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ANXA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR