Anti-GABRA4 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-783EA 553 EUR
PRSI46-783
Anti-GABRA4 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:16000. Western Blot: Approximately 70 kDa band observed in human brain (cerebellum and frontal cortex) lysates (calculated MW of 61.6 kDa according to NP_000800.2). Recommended concentration: 0.1-0.3 μg/mL. Immunohistochemistry: In paraffin embedded human cortex shows patchy staining near or at the neuronal cell membrane. Recommended concentration, 2-4 μg/mL.

GABRA4 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

GABRA4 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GABRA4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR