Anti-EPM2A Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-781EA 553 EUR
PRSI46-781
Anti-EPM2A Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:16,000. Western Blot: Approximately 38 kDa band observed in human cerebellum, mouse brain and rat brain lysates. Recommended concentration: 0.1-0.3 μg/mL. Immunohistochemistry: In paraffin embedded human cerebellum shows pixulate staining in the cells of the granular layer. Recommended concentration, 2.5-3.8 μg/mL.

EPM2A antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

EPM2A antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: EPM2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR