Anti-DES Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-777EA 420 EUR
PRSI46-777
Anti-DES Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:2,000. Western Blot: Approximately 55 kDa band observed in human skeletal muscle lysates (calculated MW of 53.5 kDa according to NP_001918.3). Recommended concentration: 0.1-0.3 μg/mL. Immunohistochemistry: In paraffin embedded human prostate shows cytoplasm and cell membrane staining in stromal cells. Recommended concentration, 3-6 μg/mL.

Desmin antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Desmin antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: DES
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR