Anti-ORAI1 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-765EA 420 EUR
PRSI46-765
Anti-ORAI1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:64,000. Western Blot: Approximately 50-55 kDa band observed in human liver and human placenta lysates (calculated MW of 32.7 kDa according to NP_116179.2). The observed molecular weight corresponds to earlier findings in literature with different antibodies (Li et al,,J Biol Chem. 2007 Oct 5;282 (40) :29448-56.; PMID: 17702753). Recommended concentration: 0.03-0.1 μg/mL.

ORAI1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

ORAI1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ORAI1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR