Anti-APOL3 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-763EA 553 EUR
PRSI46-763
Anti-APOL3 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:1,000. Western Blot: Approximately 40 kDa band observed in lysates of cell lines DAUDI and MOLT4 (calculated MW of 44.3 kDa according to NP_663615.1). Recommended concentration: 1-3 μg/mL.

APOL3 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

APOL3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: APOL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR