Anti-PCSK6 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-742EA 420 EUR
PRSI46-742
Anti-PCSK6 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:8,000. Western Blot: Approximately 70 kDa band in human, mouse and rat liver lysates after 1 μg/mL antibody staining. The 70 kDa band was successfully blocked by incubation with the immunizing peptide.

PACE4 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

PACE4 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PCSK6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR