Anti-ABCE1 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-735EA 435 EUR
PRSI46-735
Anti-ABCE1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 80 kDa band in human spleen lysates after 0.2 μg/mL antibody staining.3 kDa according to NP_002931.2. The ~80 kDa band was successfully blocked by incubation with the immunizing peptide.

RNase L antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

RNase L antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ABCE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR