Anti-CERS3 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-723EA 420 EUR
PRSI46-723
Anti-CERS3 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 45 kDa band observed in rat testis lysates (calculated MW of 46.3 kDa according to human NP_849164.2 and of 47.2 kDa according to rat NP_001121033.1). Recommended concentration: 2-3 μg/mL.

LASS3 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

LASS3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CERS3
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR