Anti-PPP2R3A Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-701EA 420 EUR
PRSI46-701
Anti-PPP2R3A Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:16,000. Western Blot: Approximately 80 kDa band in human muscle lysates after 1 μg/mL antibody staining. Please note that currently we cannot find an explanation in the literature

PPP2R3A antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

PPP2R3A antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PPP2R3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR