Anti-MARCH7 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-686EA 420 EUR
PRSI46-686
Anti-MARCH7 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:4,000. Western Blot: Approximately 125 kDa band in human spleen and tonsil lysates after 1 μg/mL antibody staining. Please note that currently we cannot find an explanation in the

MARCH7 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

MARCH7 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MARCH7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR