Anti-MARCH1 Antibody

Lieferant: ProSci Inc.
PRSI46-684EA 345 EUR
PRSI46-684
Anti-MARCH1 Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32,000.

MARCH1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

MARCH1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MARCH1
Clonality:
Clone:
Conjugation:
Epitope:
Host:
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR