Anti-KCNB1 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-678EA 345 EUR
PRSI46-678
Anti-KCNB1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:4,000. Western Blot: Preliminary experiments gave a 100 kDa band and additional stronger bands at approximately 50 kDa and 38 kDa in human cerebellum and heart lysates after 0.5 μg/mL antibody staining. Ple

KCNB1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

KCNB1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: KCNB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR