Anti-GRIN2D Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-670EA 435 EUR
PRSI46-670
Anti-GRIN2D Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 80 kDa band in human cerebellum and in mouse brain and rat brain lysates after 0.02 μg/mL antibody staining. Please note that currently

GRIN2D antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

GRIN2D antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GRIN2D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR