Anti-ESRRG Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-655EA 553 EUR
PRSI46-655
Anti-ESRRG Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:16,000. Western Blot: Approximately 50 kDa band observed in human cerebellum and breast lysates (calculated MW of 51.3 kDa according to NP_NP_001429.2). In transfected HEK293 transiently expressing ESRRG a band of approximately 52 kDa is observed. This band is not observed in the non-transfected HEK293. Recommended concentration: 0.2-0.6 μg/mL.

ESRRG antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

ESRRG antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ESRRG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR