Anti-DNMT1 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-652EA 420 EUR
PRSI46-652
Anti-DNMT1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 170 kDa band observed in Jurkat lysate (calculated MW of 183 kDa according to NP_001370.1). Recommended concentration: 0.5-1.5 μg/mL. Immunohistochemistry: In paraffin embedded human kidney shows nuclear staining in PCT. Recommended concentration, 3-6 μg/mL.

DNMT1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

DNMT1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: DNMT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR