Anti-DACH2 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-650EA 553 EUR
PRSI46-650
Anti-DACH2 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:64,000. Western Blot: Approximately 65-70 kDa band observed in lysates of cell line NIH3T3 (calculated MW of 65.3 kDa according to human NP_444511.1 and of 68.6 kDa according to mouse NP_291083.1).

DACH2 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

DACH2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: DACH2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR