Anti-CLEC16A Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-644EA 420 EUR
PRSI46-644
Anti-CLEC16A Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:16,000. Western Blot: Approximately 70 kDa band in rat testis lysates after 0.5 μg/mL antibody staining. Please note that currently we cannot find an explanation in the literature

CLEC16A antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

CLEC16A antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CLEC16A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR