Anti-ASS1 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-626EA 420 EUR
PRSI46-626
Anti-ASS1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:64000. Western Blot: Approximately 45 kDa band observed in human kidney lysates (calculated MW of 46.5 kDa according to NP_000041.2 and NP_446464.1). Recommended concentration: 0.01-0.03 μg/mL.

Argininosuccinate Synthetase 1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Argininosuccinate Synthetase 1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ASS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR