Anti-APOBEC4 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-625EA 345 EUR
PRSI46-625
Anti-APOBEC4 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 48 kDa band in human bone marrow lysates after 0.03 μg/mL antibody staining. Please note that currently we cannot find an explanation in the lit

APOBEC4 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

APOBEC4 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: APOBEC4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR