Anti-APOBEC2 Goat Polyclonal Antibody

Lieferant: ProSci Inc.
PRSI46-624EA 420 EUR
PRSI46-624
Anti-APOBEC2 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:1,000. Western Blot: Approximately 26 kDa band observed in human heart and muscle lysates (calculated MW of 25.7 kDa according to NP_006780.1). Recommended concentration: 1-3 μg/mL. an additional band of 90 kDa was consistently observed, however this band was not blocked by the immunizing peptide and it is therefore a non-specific signal.

APOBEC2 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

APOBEC2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: APOBEC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR