Anti-GAGED2 Goat Polyclonal Antibody

PRSI46-605EA 345 EUR
PRSI46-605
Anti-GAGED2 Goat Polyclonal Antibody
ELISA: Antibody detection limit dilution 1:2,000. Western Blot: preliminary experiments in human testis lysates and in lysates of different human epithelial cell lines gave no specific signal but low background (at antibody concentration up to 1 μg/mL).

XAGE1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

XAGE1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GAGED2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR